(±)-1,2-Propanediol, EMPLURA®, Supelco®
Supplier: MilliporeSigma
Synonyms:
Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
M-Clarity™ quality level = MQ200
8.22324.1000
8.22324.5000
EM8.22324.1000CS
861
USD
EM8.22324.1000
EM8.22324.5000
(±)-1,2-Propanediol, EMPLURA®, Supelco®
(±)-1,2-Propanediol
Formula:
CH₃CH(OH)CH₂OH MW: 76.1 g/mol Boiling Pt: 187.6 °C (1013 hPa) Melting Pt: –59 °C Density: 1.04 g/cm³ (20 °C) Flash Pt: 99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064272 CAS Number: 57-55-6 EINECS: 200-338-0 Merck Index: 13,07947 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...