(±)-α-Lipoic acid ≥98.0% (dry basis)
Supplier: Thermo Fisher Scientific
|
Warning
|
J18505-18
J18505-14
J18505-22
101170-522EA
400.62
USD
101170-522
101171-002
101172-678
(±)-α-Lipoic acid ≥98.0% (dry basis)
(±)-α-Lipoic acid
Formula:
C₈H₁₄O₂S₂ Boiling Pt: 160…165 °C Melting Pt: 59…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005474 CAS Number: 1077-28-7 EINECS: 214-071-2 Merck Index: 14,9326 |
Specification Test Results
Assay (Dry Basis) | ≥98.0% |
Identity | Passes Test |
Loss on Drying | ≤0.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...