Lithium dodecyl sulphate 98+% (by acidimetry)
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Dodecyl sulphuric acid lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulphate lithium salt
J32816-22
J32816-14
J32816-A1
101375-330EA
281.08
USD
101375-330
101174-788
103728-124
Lithium dodecyl sulphate 98+% (by acidimetry)
Lithium dodecyl sulphate
Formula:
C₁₂H₂₅LiO₄S Storage Temperature: Ambient |
MDL Number:
MFCD00007467 CAS Number: 2044-56-6 EINECS: 218-058-2 UN: 1325 ADR: 4.1,III |
Specification Test Results
Form | White crystalline powder |
Assay (unspecified) | ≥98% (acidity) |
Assay (unspecified) Absorbance (230 nm, 3%, 1 cm) | ≤ 0.2 |
Assay (unspecified) Absorbance (280 nm, 3%, 1 cm) | ≤ 0.1 |
Assay (GC) (C12) | ≥98% |
Chloride content | ≤ 0.01% |
Loss on Drying | ≤ 1% |
Trace Metal: Copper (Cu) | ≤ 5 ppm |
Trace Metal: Iron (Fe) | ≤ 1 ppm |
Trace Metal: Lead (Pb) | ≤ 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...