Chemical Preservative Glass Ampoules, Thermo Scientific
Supplier: Thermo Scientific
ACN-.5 ACH-1 ACN-5 ACN-5-1 ACH-10-1
89040-022CS 172.22 USD
89040-022 EPACH1 EPACN5 EPACN51 EPACH-10-1
Chemical Preservative Glass Ampoules, Thermo Scientific
Ampules
Timesaving ready to use pre-mixed chemical preservative glass ampules.
  • Pre-formulated and aliquoted glass ampoules are ready for field use
  • Ampule breaks at the pre-scored neck area by using the provided “breaker” tool
Ampules are machine-welded closed after filling for a consistent, leak-tight seal.
Envase: Packaged and shipped in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification regulations.