Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥98.5% (dry basis), ultrapure

Supplier: Thermo Fisher Scientific
Warning

J12112-36 J12112-A1 J12112-22
101174-242EA 405 USD
101174-242 101174-240 101172-256
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥98.5% (dry basis), ultrapure
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS)
Formula: C₈H₁₉NO₅
Melting Pt: 102 - 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00002853
CAS Number: 6976-37-0
EINECS: 230-237-7

Order Now

Specification Test Results

Assay Absorbance (280 nm, 1 M, 1 cm) ≤0.4
Assay (dry basis) ≥98.5%
Identification (FTIR) Conforms
Loss on Drying ≤0.5%
pKa (20 °C) 6.50 ±0.15
Iron (Fe) ≤30 ppm
Lead (Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR