Kanamycin sulfate, Ultrapure
Supplier: Thermo Fisher Scientific
|
Danger
|
A selectable marker for cosmid vectors used in transfection of mammalian cells.
It binds to ribosomal components and inhibits protein synthesis. Kanamycin is inactivated by an aminoglycoside phospho transferase (MW 25,000) in the periplasmic space.
Formula:
C₁₈H₃₈N₄O₁₅S Storage Temperature: Refrigerator |
MDL Number:
MFCD00070253 CAS Number: 25389-94-0 EINECS: 246-933-9 Merck Index: 14,5281 |
Specification Test Results
Identity (A, B, C) | Passes Test |
Potency | ≥750 µg of C₁₈H₃₆N₄O₁₁/mg powder |
Residue on Ignition | ≤1.0% |
Impurity Content Organic Impurities | Meets the Requirements |
Loss on Drying | ≤4.0% |
pH (1%, H₂O) | 6.5 - 8.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...