L(+)-Cysteine hydrochloride monohydrate 98.5-101.5% (dry basis)
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(R)-(+)-Cysteine hydrochloride monohydrate, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, L-Cysteine HCI Monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, Monohydrate, Cysteine Hydrochloride monohydrate
J14035-A1
J14035-22
J14035-A7
J14035-14
101174-990EA
396.28
USD
101174-990
101174-992
101171-578
103728-078
L(+)-Cysteine hydrochloride monohydrate 98.5-101.5% (dry basis)
L(+)-Cysteine hydrochloride monohydrate
Formula:
HSCH₂CH(NH₂)COOH·HCl·H₂O MW: 175.64 g/mol Melting Pt: ∼ 175 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00065606 CAS Number: 7048-04-6 EINECS: 200-157-7 Merck Index: 13,02810 |
Specification Test Results
Form | White crystalline powder |
Assay (unspecified) | 98.5 to 101.5% (dry basis) |
Identification (FTIR) | Conforms |
Residual halide: Chlorine (Cl) | 19.8 - 20.8% |
Residue on ignition | ≤ 0.1% |
Impurity content: Sulfate (SO4) | ≤ 0.03% |
Loss on Drying | 8.0 to 12.0% |
Optical Rotation | +5.7° to 6.8° (c=8 in in 6N HCl, dry basis) |
Thin Layer Chromatography | Homogeneous |
Trace Metal: Arsenic (As) | ≤ 1.5 ppm |
Trace Metal: Heavy Metals (Pb) | ≤ 10 ppm |
Trace Metal: Iron (Fe) | ≤ 10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...