L(+)-Leucine 98.5-100.5% (dry basis), Ultrapure

Supplier: Thermo Fisher Scientific

Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine

J18280-A1 J18280-22 J18280-A7
101171-704EA 345.18 USD
101171-704 101172-666 101171-706
L(+)-Leucine 98.5-100.5% (dry basis), Ultrapure
L(+)-Leucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: >300 °C
Density: 1.165 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002617
CAS Number: 61-90-5
EINECS: 200-522-0
Merck Index: 14,05451

Order Now

Specification Test Results

Assay (Dry Basis) 98.5 - 101.5%
Identification (FTIR) Conforms
Residue on Ignition ≤0.4%
Loss on Drying ≤0.2%
Optical Rotation +14.9 - 17.3° (c=4 in 6 N HCl Dry Basis)
Individual Impurities (Thin Layer Chromatography) ≤0.5%
Total Impurities (Thin Layer Chromatography) ≤2.0%
Trace Metal, Heavy Metals (Pb) ≤15 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR