L(+)-Histidine monohydrochloride monohydrate 98+%

Supplier: Thermo Fisher Scientific

Synonyms: L-Histidine monohydrochloride monohydrate, (2S)-(+)-Histidine monohydrochloride monohydrate, L-Histidine hydrochloride monohydrate, (S)-(+)-Histidine monohydrochloride monohydrate, His HCl monohydrate, H-His-OH.HCl.H2O, H-His-OH·HCl·H₂O, L-3-Imidazol-4-ylalanine monohydrochloride monohydrate, L-Histidine hydrochloride hydrate

J17080-22 J17080-A1
101175-554EA 83 USD
101175-554 101175-056
L(+)-Histidine monohydrochloride monohydrate 98+%
L(+)-Histidine monohydrochloride monohydrate
Formula: C₆H₉N₃O₂·HCl·H₂O
MW: 209.63 g/mol
Melting Pt: ∼ 240 °C (dec.)
Density: 1.485 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151027
CAS Number: 5934-29-2
EINECS: 211-438-9
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561d2381f930afe618f78d680790ea9a43f6596c&vid=8aacf6dc4ef5382410cb743498272b48c3c36999&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (unspecified) - Ammonium (NH₄) ≤0.02%
Assay (unspecified) ≥98.0%
Identification (FTIR) Conforms
Residue on Ignition ≤0.10%
Impurity Content Sulphate (SO₄) ≤0.03%
Loss on Drying ≤0.3%
Optical Rotation (c= in 6 N HCl) +9.2 to 10.6°
Thin Layer Chromatography Homogeneous
Arsenic (As₂O₃) ≤1 ppm
Iron (Fe) ≤30 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR