L(+)-Glutamine ≥98.5%, Ultrapure

Supplier: Thermo Fisher Scientific

Synonyms: Q(S)-2,5-Diamino-5-oxopentanoic acidL-GlutamineLevoglutamide(S)-(+)-GlutamineGlnH-Gln-OH

J16285-30 J16285-A1 J16285-36 J16285-22
101170-462EA 169.48 USD
101170-462 101172-558 101175-036 101172-556
L(+)-Glutamine ≥98.5%, Ultrapure
L(+)-Glutamine
Formula: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Melting Pt: 185…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS Number: 56-85-9
EINECS: 200-292-1
Merck Index: 14,4471
https://pr.vwr.com/stibo/web/std.lang.all/98/28/2299828.jpg

Order Now

Specification Test Results

Assay (unspecified) ≥98.5%
Chloride Content ≤0.021%
Identification (FTIR) Conforms
Residue on ignition ≤0.1%
Impurity content, Sulfate (SO₄) ≤0.028%
Loss on Drying ≤0.2%
Optical Rotation 6.3° - 7.3° (c=4 in H₂O)
Trace Metal, Arsenic (As) ≤3 ppm
Trace Metal, Heavy Metals (Pb) ≤10 ppm
Trace Metal, Iron (Fe) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR