β-(N-Morpholino)ethanesufonic acid (MES) monohydrate ≥98.5% (dry basis), Ultrapure
Supplier: Thermo Fisher Scientific
Synonyms:
MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
J18886-22
J18886-A1
101175-102EA
157.02
USD
101175-102
101170-536
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate ≥98.5% (dry basis), Ultrapure
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Formula:
C₆H₁₃NO₄S·1H₂O MW: 213.25 g/mol Melting Pt: ? 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149409 CAS Number: 145224-94-8 EINECS: 224-632-3 Merck Index: 13,05929 |
Specification Test Results
Assay (unspecified) - Absorbance (250 nm, 1 M, 1 cm) | ≤0.1 |
Assay (unspecified) - (dry basis) | ≥98.5% |
Identification (FTIR) | Conforms |
Water Content (Karl Fischer Titration) | ≤11% |
pKa (20 °C) | 6.15 ±0.15 |
Iron (Fe) | ≤ 30 ppm |
Lead (Pb) | ≤ 10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...