Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99,8-100,1 Dry_basis, ultrapure Molecular_Biology

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

J75825-36 J75825-A1 J75825-A7 J75825-A9
101174-856EA 112.03 USD
101174-856 101170-654 101173-310 101375-578
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99,8-100,1 Dry_basis, ultrapure Molecular_Biology
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 357
Melting Pt: 171…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 14,9772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (unspecified) - Dry Basis 99.8 - 100.1%
Assay (unspecified) - Absorbance (250 nm, 1 M, 1 cm) ≤0.1
Assay (unspecified) - Absorbance (290 nm, 40%, 1 cm) ≤0.2
Identification (FTIR) Conforms
Residue on Ignition ≤0.1%
Insoluble Material ≤0.005%
Water Content (Karl Fischer Titration) ≤1%
Loss on Drying ≤1%
pH (5%, H₂O) 10.0 - 11.5
pKa (20 °C) 8.30 ±0.15
Arsenic (As) ≤1 ppm
Barium (Ba) ≤1 ppm
Cadmium (Cd) ≤1 ppm
Calcium (Ca) ≤1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤1 ppm
Magnesium (Mg) ≤1 ppm
Manganese (Mn) ≤1 ppm
Zinc (Zn) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR