Microflex® E-Grip® MAX L92 Powder-Free Latex Examination Gloves
Supplier: Ansell Healthcare
L920 L921 L923 L924 L922
10118-992BX 34.22 USD
10118-992 10118-994 10118-998 10119-000 10159-882
Microflex® E-Grip® MAX L92 Powder-Free Latex Examination Gloves
Gloves
Microflex® E-Grip® MAX is an examination glove with a thick design for enhanced durability.
  • Ambidextrous
  • Nonsterile
  • Powder-free
  • Textured fingers
Fully textured fingers offer enhanced gripping power in every situation. Gloves are natural in color. They also feature textured fingers and beaded cuffs.
These ultra-durable exam gloves made of a soft, comfortable latex have added thickness for extra protection. Because the glove is double chlorinated, it is easy to don and doff.
Certificaciones: 510K approved for medical use.