Agar Noble agar, Ultrapure

Supplier: Thermo Fisher Scientific

Synonyms: Agar Agar

J10907-36 J10907-22 J10907-A1
101170-362EA 295.34 USD
101170-362 101171-498 101172-166
Agar Noble agar, Ultrapure
Agar

Gelling agent essentially free of impurities. Used mainly in biochemistry and molecular biology procedures wherever increased purity is required.

Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Absorbance (450 nm, 1.5%) ≤0.150
Assay Gel Point (1.5%) 32 - 37.5 °C
Assay Gel Strength (1.5%) ≥700 gm/cm2
Assay Gel pH (1.5%) 5.0 - 7.0
Ash ≤1.6%
Comment EEO (-Mr): ≤0.450 (pH 8.4)
Resistance (1% suspension) ≥20,000 Ohms
Residual water ≤10%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR