Oblong Bottles, Wide Mouth, HDPE, with Lined Closure, Thermo Scientific
Supplier: Thermo Scientific
316-0125 316-0250BPC 316-0500BPC 316-1000 316-1000BPC
IR316-0125CS 197.24 USD
IR316-0125 IR316-0125BPC IR316-0250 IR316-0250BPC IR316-0500 IR316-0500BPC IR316-1000 IR316-1000BPC
Oblong Bottles, Wide Mouth, HDPE, with Lined Closure, Thermo Scientific
Bottles Plastic Bottles
Save valuable space and stabilize samples within shipping containers.
  • Available bulk-packed for added economy
Each container is individually barcoded with lot number and unique container number. Analysis labels are included unattached for optional use. Packaging cartons are sealed with security tape to provide tamper evidence.
The space-saving oblong shape and flat side panels allow bottles to be packed tightly into coolers. Wide mouth HDPE oblong containers are an excellent general-purpose sample container for liquid sampling in inorganic analyte testing. The wide mouth feature makes this bottle ideal for sampling more viscous liquid samples like industrial wastes and sludges.
Certificaciones: Certified containers meet or exceed EPA performance-based standards for metals, cyanide, and fluoride. Manufactured under an ISO certified quality management system.