Modesty Panel for C-Leg Bench, Production Basics

Supplier: Production Basics
8712 8705 8709 8715
11005-472EA 127.7 USD
11005-472 11005-468 11005-470 11005-474
Modesty Panel for C-Leg Bench, Production Basics
Furniture Tables and Workbenches
Steel panel mounts to frame below the worksurface for privacy.

  • For use with any Production Basics C-Leg lab bench

Color matches frame finish of workstation when ordered together.

Color matches frame finish of workstation when ordered together.

Información para pedidos: Specify a frame color for panel otherwise will be default blue frame. Frame available in light neutral/almond, blue, white, gray and black. Accessory is ready to mount on a C-Leg bench, sold separately.

Información de suministro: Includes (1) modesty panel, (1) parts and hardware packet.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR