Anti-CD2 Rat Antibody (PE (Phycoerythrin)) [clone: RM2-5]

Supplier: Tonbo Biosciences
50-0021-U025 50-0021-U100
10141-190EA 52.8 USD
10141-190 10141-192
Anti-CD2 Rat Antibody (PE (Phycoerythrin)) [clone: RM2-5]
Antibodies
The RM2-5 antibody reacts with mouse CD2, an approximately 50 kDa glycoprotein, and a member of the Ig superfamily. CD2, also known as LFA-2, is a receptor for CD48 in the mouse and is expressed on the cell surface of all mouse lymphocytes. CD2 mediates adhesion between cells and is involved in T cell activation.

RM2-5 is reported to block CD2-mediated cell:cell adhesion. Please choose the appropriate format for each application.

Type: Primary
Antigen: CD2
Clonality: 0
Clone: RM2-5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR