Anti-CD44 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: IM7]

Supplier: Tonbo Biosciences
35-0441-U025 35-0441-U100 35-0441-U500
10050-894EA 30.8 USD
10050-894 10050-896 10050-898
Anti-CD44 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: IM7]
Antibodies
The IM7 antibody recognizes CD44, a ubiquitously expressed cell surface receptor which is important for extracellular matrix organization, cell-cell and cell-matrix adhesion and migration. CD44 may be expressed in a number of different isoforms (splice variants) from the most typical or “standard” form, known as CD44s, to variants designated CD44v, e.g. CD44v1 or CD44v6. These receptors interact with several ligands, but most often associate with an extracellular matrix component hyaluronate, through which it mediates adhesion.

The IM7 antibody may be used for detection of all isoforms of CD44, as it recognizes constant epitopes near the extracellular proximal domain. (Xu et al, 2002, J. Leukoc. Biol. 72:1133-1141). It has been reported to be cross-reactive with many non-human species including Baboon, Chimpanzee, Cynomolgus, Rhesus, Horse, Cow, Pig, Dog and Cat CD44.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR