EZ-CFU™, Microbiologics
Supplier: MICROBIOLOGICS, INC.
0392C 0998C 0486C 0443C 0487C 0317C 0306C 0366C 0483C 0872C 0871C 0688C 0813C 0524C 0890C 0902C 0421C 0363C 0360C 0827C 0485C 0371C 0385C 0687C 0353C 0484C
10142-660EA 1005.61 USD
10142-660 10142-662 10142-664 10142-668 10142-670 10142-672 10142-674 10142-676 10142-682 10142-684 10142-686 10142-688 10142-690 10142-692 10142-694 10142-696 10142-698 10142-700 10142-702 10142-704 10142-706 10142-708 10142-710 10142-724 10142-726 10142-728
EZ-CFU™, Microbiologics
Controls and Calibration Verification Materials
EZ-CFU™ is used to perform Growth Promotion Tests of culture media with ease
EZ-CFU™ kits include 20 lyophilized pellets of a single quantitative QC microorganism, 10 vials of hydrating fluid (2 mL per vial) and Peel-Off Certificate of Assay. Quantitative testing saves time and reduces labor by delivering a specific number of colony forming units (CFU).
EZ-CFU™ lyophilized QC microorganism preparations deliver 10-100 CFU per 0.1 mL when processed as directed. A one log dilution in pH 7.2 phosphate buffer is required. Following the dilution step, EZ-CFU™ provides over 90 inocula using the same suspension.
Información para pedidos: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the product page. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.