Anti-CD8A Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: 53-6.7]

Supplier: Tonbo Biosciences
65-0081-U025 65-0081-U100
10051-560EA 59.4 USD
10051-560 10051-562
Anti-CD8A Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: 53-6.7]
Antibodies
The 53-6.7 antibody reacts with the 32-34 kDa alpha subunit of mouse CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor in antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development,More Product Information
Order Now

SPECIFICATIONS

Antigen Symbol CD8A
Antigen Name CD8 antigen, alpha chain
Environmentally Preferable
Antigen Synonyms p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
Antibody Type Primary
Clonality Monoclonal
Conjugation PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Clone 53-6.7
Reactivity Mouse
Host Rat
Gene ID 12525
Isotype IgG2a
Flow Cytometry Yes
Neutralisation Yes
Storage Temperature Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
Tested Applications Flow Cytometry

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR