1,1,2,2-Tetrachloroethane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.43948.0250
8.43948.1000
EM8.43948.0250CS
550
USD
EM8.43948.0250
EM8.43948.1000
1,1,2,2-Tetrachloroethane for synthesis, Sigma-Aldrich®
1,1,2,2-Tetrachloroethane
Formula:
Cl₂HCCHCl₂ MW: 167.85 g/mol |
MDL Number:
MFCD00000848 CAS Number: 79-34-5 UN: 1702 ADR: 6.1,II |
Specification Test Results
Solubility | 2830 g/l (25°C) |
Melting Point | -42.5 °C |
Vapor Pressure | 6.4 hPa (20 °C) |
Density | 1.59 g/cm3 (25 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...