High Temperature Clamps, NW Flange, Ace Glass Incorporated
Supplier: Ace Glass
12189-02 12189-04 12189-06
10099-152EA 153.04 USD
10099-152 10099-154 10099-156
High Temperature Clamps, NW Flange, Ace Glass Incorporated
Clamps General Purpose Clamps
High temperature NW flange clamps
Temperature range -200 to +200°C vacuum to 10E-7 and pressure up to 10 bar. Clamps are used in food, dairy, beverage, pharmaceutical, and biotech applications. Fully sterilizable.