Ammonium iron (III) citrate, Reagent Grade

Supplier: VWR

Synonyms: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate

0846-100G 0846-500G
89230-024EA 81.45 USD
89230-024 89230-026
Ammonium iron (III) citrate, Reagent Grade
Ammonium iron (III) citrate
Formula: C₆H₅+4y)Fe(x)N(y)O₇
Storage Temperature: Ambient
MDL Number: MFCD00013099
CAS Number: 1185-57-5
EINECS: 214-686-6

Order Now

Specification Test Results

Arsenic <= 0.0004%
Iron (III) 20.5% - 22.5%
Lead <= 0.003%
Solubility (5.0%, Water) Pass
Zinc <= 0.1%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR