Methyl orange ACS

Supplier: VWR
Danger

0596-100G 0596-25G
89399-850CS 823.44 USD
89399-850 89399-852
Methyl orange ACS
Methyl orange
Formula: C₁₄H₁₄N₃NaO₃S
Boiling Pt: ∼90 °C (1013 hPa)
Melting Pt: >300 °C
Density: ~ 1 g/cm³ (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
UN: 3143
ADR: 6.1,III
Merck Index: 13,06127

Order Now

Specification Test Results

Clarity of Solution Pass
Visual Transition Interval (pH 3.2-4.4) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR