Fritted Support Assemblies, 47 mm, Chemglass
Supplier: Chemglass
CG-1420-01 CG-1420-12 CG-1424-01 CG-1420-02 CG-1420-03 CG-1420-04 CG-1420-05 CG-1420-10
80068-654EA 598.03 USD
80068-654 80068-668 80068-678 80068-656 80068-658 80068-660 80068-662 80068-664
Fritted Support Assemblies, 47 mm, Chemglass
Filters Membrane Filter Units
Glassware system for vacuum filtration using 47 mm membranes.
Assembly is designed for use with vacuum manifolds or optional plastic coated filtering flask. Support base has a sealed-in coarse porosity fritted disc. Complete assembly includes a 300 ml graduated funnel, aluminum clamp, support base and silicone stopper.