Anti-WT1 Mouse Monoclonal Antibody [clone: 6F-H2]

Supplier: Diagnostic Biosystems

MOB437 MOB437-05 PDM177
10065-068EA 839.29 USD
10065-068 10065-070 10065-088
Anti-WT1 Mouse Monoclonal Antibody [clone: 6F-H2]
Antibodies
Wilms tumor-1 (WT-1), is a gene involved in the induction of Wilms tumor, a pediatric renal malignancy. Wilms tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed in fetal kidney and mesothelium, and its expression has been suggested as a marker for Wilms tumor and mesothelioma.

Type:
Antigen: WT1
Clonality: Monoclonal
Clone: 6F-H2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR