Anti-DOG 1 Mouse Monoclonal Antibody [clone: DOG 1.1]

Supplier: Diagnostic Biosystems

MOB466 MOB466-05 PDM170
10064-950EA 639.62 USD
10064-950 10064-952 10064-992
Anti-DOG 1 Mouse Monoclonal Antibody [clone: DOG 1.1]
Antibodies
DOG1 is expressed ubiquitously in gastrointestinal stromal tumors irrespective of c-kit or PDGFR alpha mutation status. Reactivity for DOG1 may aid in the diagnosis of gastrointestinal stromal tumors, including PDGFRA mutants that fail to express c-kit antigen, and lead to appropriate treatment with imatinib mesylate, an inhibitor of the c-kit tyrosine kinase.

Type:
Antigen: DOG 1
Clonality: Monoclonal
Clone: DOG 1.1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR