UnoVue™ Mouse/Rabbit AP Detection Reagent, Detection Systems, Diagnostic BioSystems
Supplier: Diagnostic Biosystems
MRU-AP100 MRU-AP1000
10065-128EA 410.55 USD
10065-128 10065-130
UnoVue™ Mouse/Rabbit AP Detection Reagent, Detection Systems, Diagnostic BioSystems
Assays Immunological Assays
UnoVue Mouse/Rabbit AP Detection Reagent is a non-biotin one-step detection reagent suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and cryostat sections.
The UnoVue detection reagent may also be used with blood smears, cytosmears, and cell preparations.
Order Now
SPECIFICATIONS
Environmentally Preferable