PolyVue Plus™ Mouse/Rabbit HRP/DAB Detection System, Diagnostic BioSystems
Supplier: Diagnostic Biosystems
PVP1000D PVP100D
10065-148EA 2577.72 USD
10065-148 10065-150
PolyVue Plus™ Mouse/Rabbit HRP/DAB Detection System, Diagnostic BioSystems
Assays Immunological Assays
The PolyVue Plus™ Mouse/Rabbit/HRP/DAB detection system is a non-biotin, two-step detection system suitable for labeling antigens in formalin-fixed paraffin-embedded tissues and cryostat sections.
The PolyVue Plus™ detection system may also be used with blood smears, cytosmears, and cell preparations. This system is developed by directly labeling immunoglobulins with enzymes using a proprietary tandem hyperlabelling technology. This ensures consistent and reproducible immunostaining for all types of nuclear, cytoplasmic and membrane antigens in different types of tissues with significantly lower background than detection systems using biotin and avidin conjugates.
Información para pedidos: The kit includes tissue primer, background blocker, PolyVue plus mouse/rabbit HRP enhancer, PolyVue plus mouse/rabbit HRP label, stable DAB plus buffer, stable DAB plus chromogen, and an empty mixing bottle.