Anti-Microphthalmia Transcription Factor Mouse Monoclonal Antibody [clone: D5]

Supplier: Diagnostic Biosystems

PDM168 MOB462 MOB462-05
10064-988EA 464.64 USD
10064-988 10065-072 10065-074
Anti-Microphthalmia Transcription Factor Mouse Monoclonal Antibody [clone: D5]
Antibodies
This antibody reacts with a 52-56 kDa protein known as microphthalmia (Mi). This antibody reacts with both melanocytic and non-melanocytic isoforms of Mi. Mi is a basic helix-loop-helix-leucine zipper (b-HLH-ZIP) transcription factor implicated in pigmentation, mast cells and bone development. There are two known isoforms of Mi differing by 66 amino acids at the amino terminus.

Type:
Antigen: Microphthalmia Transcription Factor
Clonality: Monoclonal
Clone: D5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR