Food Hoes, Remco Products
Supplier: Remco Products
29002 29003 29004 29005 29006
76007-108EA 38.03 USD
76007-108 76007-110 76007-112 76007-114 76007-116
Food Hoes, Remco Products
Scrapers
Here is a unique scraper that is designed for both push and pull applications
It has the strength to move almost any type of product without breaking under pressure. Lightweight and long-lasting, its flat and arched edges make it the perfect tool for use on straight or curved surfaces. Nylon material.
Certificaciones: FDA Compliant Raw Material.