Anti-ErpN Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-C10S 200-401-C10
RL200-401-C10SEA 151.49 USD
RL200-401-C10S RL200-401-C10
Anti-ErpN Rabbit Polyclonal Antibody
Antibodies
Anti-ErpN/OspE antibody has been tested in Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band at 17.1 kDa in size corresponding to ErpN/OspE by Western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: >1:5000; Western Blot: 1:1000;

Type: Primary
Antigen: ErpN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgdorferi
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR