Anti-RELA Mouse Monoclonal Antibody [clone: 27F9.G4]

Supplier: Rockland Immunochemical
200-301-065 200-301-065S
RL200-301-065EA 565.22 USD
RL200-301-065 RL200-301-065S
Anti-RELA Mouse Monoclonal Antibody [clone: 27F9.G4]
Antibodies
This Protein A purified mouse monoclonal antibody is directed against NFkB p65 (Rel A) and a 1:1000 dilution recognizes a 65 kD band by Western blot against HeLa whole cell lysate. Control peptide (100-4165p) is sold separately.

Recommended Dilutions: ELISA: 1:50000-1:100000; Immunofluorescence: 1:5000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:200-1:600;

Type: Primary
Antigen: RELA
Clonality: Monoclonal
Clone: 27F9.G4
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR