Anti-CD21 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Bu32]

Supplier: Biolegend
354909 354910
354909-BLEA 125.69 USD
354909-BL 354910-BL
Anti-CD21 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Bu32]
Antibodies
FITC anti-human CD21 [Bu32]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD21
Clonality: Monoclonal
Clone: Bu32
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR