Anti-CD193 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: J073E5]

Supplier: Biolegend
144505 144506
144505-BLEA 185.8 USD
144505-BL 144506-BL
Anti-CD193 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: J073E5]
Antibodies
PE anti-mouse CD193 (CCR3) [J073E5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD193
Clonality: Monoclonal
Clone: J073E5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR