Anti-CD197 (CCR7) Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 4B12]

Supplier: Biolegend
120119 120120
120119-BLEA 242.2 USD
120119-BL 120120-BL
Anti-CD197 (CCR7) Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: 4B12]
Antibodies
Brilliant Violet 421™ anti-mouse CD197 (CCR7) [4B12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD197 (CCR7)
Clonality: Monoclonal
Clone: 4B12
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR