Vented Screw Caps for Falcon® Flasks, Sterile, Corning
Supplier: Corning
354637 354639 354638
62405-152CS 99.75 USD
62405-152 62405-156 62405-154
Vented Screw Caps for Falcon® Flasks, Sterile, Corning
Closures Bottle Closures
PE with integral hydophobic 0.2 µm microporous membrane filter vent.
Vented caps minimize contamination associated with standard open incubation. Gases required for cell growth and metabolism pass freely through the vent, while microorganisms do not.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental Certification ISO 14001 - Bedford (reg. no. 14.4942)