Safranine O, Certifiable Grade
Supplier: VWR
Synonyms:
CI 50240
0574-100G
0574-25G
97061-952EA
341.36
USD
97061-952
97061-954
Safranine O, Certifiable Grade
Safranine O
Part Fleming Triple stain, a chromosome stain prepared with light green SF and in Gram stain.
Formula:
C₂₀H₁₉ClN₄ MW: 350.85 g/mol Density: 1.447 g/cm³ (22 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011759 CAS Number: 477-73-6 EINECS: 207-518-8 |
Specification Test Results
Absorbance maximum | 527 - 533 nm |
Dye Content (Anhydrous) | >=80% |
Loss on Drying | <= 4% |
P-15/P+15 | 1.1 - 1.32 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...