Broad Spectrum Negative Control
Supplier: Diagnostic Biosystems
K053-06 K053-100 K053-25
10064-846EA 74.49 USD
10064-846 10064-848 10064-850
Broad Spectrum Negative Control
Proteins and Peptides
Negative controls are used to determine the specificity of an antibody. No staining should be observed when the primary antibody is omitted.This product is specifically designed to work with rabbit, mouse and goat antibodies. The Broad Spectrum Negative Control has been titered to work with Polymer-based Secondary Systems in manual and automated IHC platforms.
  • Ready To Use
This product is specifically designed to work with rabbit, mouse and goat antibodies. The Broad Spectrum Negative Control has been titered to work with Polymer-based Secondary Systems in manual and automated IHC platforms.
Precaución: Intended Use: IVD