PermaBlack/HRP, Diagnostic BioSystems
Supplier: Diagnostic Biosystems
K062-110 K062
10064-834EA 450.9 USD
10064-834 10064-836
PermaBlack/HRP, Diagnostic BioSystems
Stains Electrophoresis Stains
PermaBlack/HRP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing horseradish peroxidase
PermaBlack/HRP can be permanently mounted to produce a sharply contrasting ebony black color that can be easily distinguished from other stains.