Hypersil™ BDS CN HPLC Columns, Thermo Scientific
Supplier: Avantor
10042-208EA 520.62 USD
10042-208 10042-222 10042-226 10042-228 10042-232 10042-234 10042-240 10042-242
Hypersil™ BDS CN HPLC Columns, Thermo Scientific
Chromatography Columns
For a robust column that can be used in both normal phase and reversed phase, use Thermo Scientific™ Hypersil BDS Cyano columns.
These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.
Información para pedidos: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).