Acclaim™ Organic Acid (OA) LC Columns, Thermo Scientific
Supplier: Thermo Scientific
062902 062903 069700 070086 070087 071987
10044-470EA 1481.27 USD
10044-470 10044-472 10044-644 10044-676 10044-678 10044-754
Acclaim™ Organic Acid (OA) LC Columns, Thermo Scientific
Chromatography Columns
For unparalleled performance in separating hydrophilic, aliphatic, and aromatic organic acids, use Thermo Scientific™ Acclaim OA reversed-phase silica columns.
These high-efficiency columns are compatible with 100% aqueous mobile phases and have excellent hydrolytic stability at low pHs for excellent selectivity in the separation of organic acids. You'll achieve high-resolution separations with fast run times and minimal sample preparation using Acclaim OA columns for the analysis and quality testing of foods, beverages, pharmaceuticals, chemical intermediates, and environmental samples.
Información para pedidos: Guard Cartridges require use of the corresponding guard holder. See Acclaim Guard Kit Holder and Coupler V-2 (Part No. 10046-750).