Supermax Robust™ Blue Nitrile Gloves
Supplier: SUPERMAX HEALTHCARE INC
93896 93897 93898 93899
75997-432CS 179.65 USD
75997-432 75997-434 75997-436 75997-438
Supermax Robust™ Blue Nitrile Gloves
Gloves
Aurelia Robust Blue Nitrile Powder Free Examination Gloves are made from our special nitrile (NBR) compound, protein free (non-latex), ambidextrous and non sterile.Each batch of gloves is air-tested to detect pinholes. Aurelia gloves conform and are tested to the highest international standards.
  • 100% Latex free
  • 5 mil thickness for improved puncture resistance
  • Micro-textured finished delivers improved grip
  • Powder free
  • Ambidextrous
  • Non sterile, Single-use
Envase: 100 pieces per box.