Diversified Biotech Richter Anti-vibration Pad
Supplier: Diversified Biotech
VIBE-1000 VIBE-2000 VIBE-3000
490007-114PK 63.38 USD
490007-114 490007-116 490007-118
Diversified Biotech Richter Anti-vibration Pad
Surface Protectors Bench Protectors
This inexpensive solution dampens vibrations generated from lab equipment.
Creates a stable surface that will keep equipment in place. Fits a variety of benchtop vortexes, shakers, rockers, and microscopes. Small, lightweight design makes transportation between benches and labs easy. Pads can be stacked for heavier units or laid side-by-side for larger equipment.
Información para pedidos: Combo Pack comes with both sizes of the Richter Pads.