Reagent Reservoirs
Supplier: Diversified Biotech
RESE-2100 RESE-5100 RESE-1100
490006-898CS 194.91 USD
490006-898 490006-906 490006-892
Reagent Reservoirs
Reagent Reservoirs
Non sterile reagent reservoirs with graduated marks on inside walls.
  • Convenient pour-off spouts on all four corners
  • A wide base provides stability and avoids spills
  • Unique "trough within a trough" maximize reagent recovery