Petri Seal™ Tape, Diversified Biotech
Supplier: Diversified Biotech
PSEAL-108B PSEAL-108C PSEAL-108R PSEAL-108W PSEAL-108Y
490006-870EA 62.09 USD
490006-870 490006-872 490006-874 490006-876 490006-878
Petri Seal™ Tape, Diversified Biotech
Tapes
Designed for microtiter plates, petri dishes, and culture plates.
  • Stretches and conforms to form a tight seal
  • Temperature resistant
  • Waterproof and chemically resistant
  • Resists abrasion and puncture
  • Blocks vapor loss and reduces the rate of gas exchange
  • Prevents the entry of contaminants
  • Leaves no adhesive residue even after two years
  • 0.5 x 108"
The seal remains flexible down to -30 °C and withstands continuous exposure from -40 to 77 °C (with intermittent exposure up to 100 °C).