Chemically Inert Polyvinyl Labels, Laser Printable, Diversified Biotech
Supplier: Diversified Biotech
CRY-0500 CRY-1000 CRY-2000 LCRY-1000 LCRY-1200 LCRY-1258 LCRY-1700 LCRY-2380
490006-720EA 53.69 USD
490006-720 490006-722 490006-724 490006-800 490006-804 490006-816 490006-822 490006-834
Chemically Inert Polyvinyl Labels, Laser Printable, Diversified Biotech
Labels
These laser labels are made of chemically inert polyvinyl labels that adhere to most plastics and other materials. These labels are rectangular, pre-cut, peel-off labels and ideal for cryogenic storage.
  • Labels are available as Cryo Babies® or Cryo Tags®
  • Chemical resistant
  • Temperature resistance: −196 to 150 °C
  • Accept any marking instrument
Unlike other tape materials that are brittle and break off of vials at low temperatures, these labels withstand conventional and cryogenic freezer storage conditions (liquid nitrogen storage) as well as boiling water baths, most organic solvents, caustic agents, and other challenges without peeling.