Cryo Labels, Laser Printable, Diversified Biotech
Supplier: Diversified Biotech
CLEAR-170 CLEAR-238
490006-694PK 163.58 USD
490006-694 490006-696
Cryo Labels, Laser Printable, Diversified Biotech
Labels
Cryo-Clear® Labels are chemically resistant, flexible labels that can withstand boiling, freezing to −196 °C, sustained humidity.
  • These labels resist most organic solvents and caustic agents
  • They adhere to most plastics, glass and metals without cracking, peeling, or degrading
  • Heat-resistant, stay-flat sheets will not jam in laser printers
  • Label sheets are top-coated to readily accept laser printing
Made of flexible vinyl label material and acrylic adhesive. This unique combination expands and contracts together with temperature fluctuations resulting in a strong adhesive bond. Labels are recommended to use with 100% virgin polypropylene tubes and avoid the use of siliconized surfaces.