Advansta Incubation Trays
Supplier: Advansta
L-07031-005 L-07033-005 L-07034-005 L-07038-005 L-07035-004 L-07039-004 L-07036-003 L-07040-003 L-07037-002 L-07041-002
490005-028PK 90.43 USD
490005-028 490005-032 490016-786 490016-788 490016-790 490016-792 490016-794 490016-796 490016-798 490016-800
Advansta Incubation Trays
Trays/Dishes/Pots Laboratory Trays/Dishes
These incubation trays are convenient and easy to use for staining and washing gels and membranes.
  • Smooth interior- Protects membranes and gels from scratches
  • Lidded- To protect experiments from dust or debries
  • Non-binding surfaces prevent antibody loss
  • Convenient- Multiple sizes available for different applications
These lidded incubation trays have a perfectly smooth interior surface, making them ideal for staining and washing electrophoresis gels and membranes. Trays are manufactured to prevent adsorption of antibodies. Three sizes are available toMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable