Advansta RapidClean™ Protein Removal Kit
Supplier: Advansta
K-01001-010
K-01001-025
R-14011-250
R-14011-B10
490005-002EA
383.27
USD
490005-002
490016-812
490016-814
490016-816
Advansta RapidClean™ Protein Removal Kit
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
RapidClean Kit includes RapidClean Resin and spin filters to aid in resin removal
- Complete removal of proteins from aqueous solutions of nucleic acids
- Protocol less than 5 minutes, start to finish
- Non-toxic alternative to phenol-chloroform extraction
- Nucleic acid recovery rates >95%
- Recovery of nucleic acids from 5 nucleotides to greater than 40 kb
- Suitable for DNA, RNA, cDNA, microRNA, oligonucleotides
RapidClean Resin arrives as a ready-to-use 10X slurry. Its blue color makes the protein pellet easy to see when pipetting. In less than 5 minutes, completely remove protein from aqueous solutions of single- and double-stranded nucleic acids.